missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Allylbenzene, 98%, Thermo Scientific Chemicals
$127.58 - $1679.37
Chemical Identifiers
CAS | 300-57-2 |
---|---|
Molecular Formula | C9H10 |
Molecular Weight (g/mol) | 118.18 |
InChI Key | HJWLCRVIBGQPNF-UHFFFAOYSA-N |
Synonym | allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene |
PubChem CID | 9309 |
IUPAC Name | prop-2-enylbenzene |
SMILES | C=CCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC102890250
|
Thermo Scientific Chemicals
102890250 |
25 mL | Glass bottle |
Each for $127.58
|
|
||||
AC102891000
|
Thermo Scientific Chemicals
102891000 |
100 mL | Glass bottle |
Each for $439.23
|
|
||||
AC102895000
|
Thermo Scientific Chemicals
102895000 |
500 mL | Glass bottle |
Each for $1,679.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
300-57-2 | |
118.18 | |
allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene | |
prop-2-enylbenzene |
C9H10 | |
HJWLCRVIBGQPNF-UHFFFAOYSA-N | |
9309 | |
C=CCC1=CC=CC=C1 |
Specifications
300-57-2 | |
0.8900g/mL | |
33°C | |
98% | |
C9H10 | |
C6H5CH2CH=CH2 | |
05,484 | |
allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene | |
HJWLCRVIBGQPNF-UHFFFAOYSA-N | |
prop-2-enylbenzene | |
9309 | |
98% | |
Allylbenzene |
Colorless to Yellow | |
156.0°C to 157.0°C | |
Authentic | |
Glass bottle | |
1.5100 to 1.5120 | |
25 mL | |
0.89 | |
Solubility in water: insoluble | |
C=CCC1=CC=CC=C1 | |
118.18 | |
118.18 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 206-095-7
RUO – Research Use Only