missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allylbenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 102891000

 View more versions of this product

Catalog No. AC102891000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Allylbenzene
Colorless to Yellow
156.0°C to 157.0°C
Authentic
Glass bottle
1.5100 to 1.5120
100 mL
0.89
Solubility in water: insoluble
C=CCC1=CC=CC=C1
118.18
118.18
Liquid
300-57-2
0.8900g/mL
33°C
98%
C9H10
C6H5CH2CH=CH2
05,484
allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene
HJWLCRVIBGQPNF-UHFFFAOYSA-N
prop-2-enylbenzene
9309
98%

Chemical Identifiers

300-57-2
118.18
allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene
prop-2-enylbenzene
C9H10
HJWLCRVIBGQPNF-UHFFFAOYSA-N
9309
C=CCC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 206-095-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only