missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 209531000

 View more versions of this product

Catalog No. AC209531000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Allylmagnesium bromide
1730-25-2,1730-25-2
18.78,84.64
0.8510g/mL
AcroSeal™ Glass bottle
H2C=CHCH2MgBr
100 mL
0.851
FEMBXICCJNZMMC-UHFFFAOYSA-M
magnesium;prop-1-ene;bromide
74413
Solution
1M solution in diethyl ether
15.37,81.22
Gray or Green-Yellow
−40°C
C3H5BrMg
MFCD00000044
3399
allylmagnesium bromide, grignard reagent, allyl magnesium bromide, sjpdabpprujp@, sjpdabpprujp@, allylmagnesiumbromid, allylmagnesiumbromid, allyl magnesiumbromide, allyl magnesiumbromide, 2-propenylmagnesium bromide, 2-propenylmagnesium bromide
Br[Mg]CC=C
145.28
145.28

Chemical Identifiers

1730-25-2
145.28
FEMBXICCJNZMMC-UHFFFAOYSA-M
74413
Br[Mg]CC=C
C3H5BrMg
MFCD00000044
allylmagnesium bromide, grignard reagent, allyl magnesium bromide, sjpdabpprujp@, sjpdabpprujp@, allylmagnesiumbromid, allylmagnesiumbromid, allyl magnesiumbromide, allyl magnesiumbromide, 2-propenylmagnesium bromide, 2-propenylmagnesium bromide
magnesium;prop-1-ene;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Harmful if swallowed.
Causes severe skin bur

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 217-046-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only