missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allylpalladium chloride dimer, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 206835000

 View more versions of this product

Catalog No. AC206835000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Allylpalladium chloride dimer
12012-95-2
100.0
56.7% min. (Pd)
C6H10Cl2Pd2
500 mg
allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
PENAXHPKEVTBLF-UHFFFAOYSA-L
chloropalladium(1+);prop-1-ene
61538
98%
98%
97.5
Green-Yellow to Yellow
Glass bottle
MFCD00044874
02,109; 05,189; 07,5; 09,132
Solubility in water: decomposes
Cl[Pd].Cl[Pd].c:c:c.c:c:c
365.89
365.84
Crystalline Powder or Crystals

Chemical Identifiers

12012-95-2
365.89
PENAXHPKEVTBLF-UHFFFAOYSA-L
61538
Cl[Pd].Cl[Pd].c:c:c.c:c:c
C6H10Cl2Pd2
MFCD00044874
allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
chloropalladium(1+);prop-1-ene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 234-579-8

RTECSNumber : RT3510000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only