Learn More
Allylpalladium chloride dimer, 98%, Thermo Scientific Chemicals
$184.72 - $1837.49
Chemical Identifiers
CAS | 12012-95-2 |
---|---|
Molecular Formula | C6H10Cl2Pd2 |
Molecular Weight (g/mol) | 365.89 |
MDL Number | MFCD00044874 |
InChI Key | PENAXHPKEVTBLF-UHFFFAOYSA-L |
Synonym | allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium |
PubChem CID | 61538 |
IUPAC Name | chloropalladium(1+);prop-1-ene |
SMILES | Cl[Pd].Cl[Pd].c:c:c.c:c:c |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC206835000
|
Thermo Scientific Chemicals
206835000 |
500 mg | Glass bottle |
Each for $184.72
|
|
||||
AC206830050
|
Thermo Scientific Chemicals
206830050 |
5 g | Glass bottle |
Each for $1,837.49
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12012-95-2 | |
365.89 | |
PENAXHPKEVTBLF-UHFFFAOYSA-L | |
61538 | |
Cl[Pd].Cl[Pd].c:c:c.c:c:c |
C6H10Cl2Pd2 | |
MFCD00044874 | |
allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium | |
chloropalladium(1+);prop-1-ene |
Specifications
12012-95-2 | |
100.0 | |
56.7% min. (Pd) | |
C6H10Cl2Pd2 | |
500 mg | |
allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium | |
PENAXHPKEVTBLF-UHFFFAOYSA-L | |
chloropalladium(1+);prop-1-ene | |
61538 | |
98% | |
Allylpalladium chloride dimer, 98% |
97.5 | |
Green-Yellow to Yellow | |
Glass bottle | |
MFCD00044874 | |
02,109; 05,189; 07,5; 09,132 | |
Solubility in water: decomposes | |
Cl[Pd].Cl[Pd].c:c:c.c:c:c | |
365.89 | |
365.84 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 234-579-8
RTECSNumber : RT3510000
RUO – Research Use Only