AlphaProTech Critical Cover™ AlphaGuard™ Lab Coats
Soft and breathable multilayer SMS material provides comfort in critical environments. Choose from elastic or knit wrist styles in blue or white colors.
Supplier:  AlphaProTech LC316326
 View more versions of this product
Catalog No. 19-130-4257
Explore available promotions
Description
Description
Soft and breathable multilayer SMS material provides comfort in critical environments. Choose from elastic or knit wrist styles in blue or white colors.
  • Soft, breathable and fluid repellent
  • Static dissipative
  • Ultra clean, low particle count
Specifications
Specifications
Controlled Environments Lab Coat
Blue
Knit
Non-sterile
Serged
97.1 cm
SMS
Snaps
Single-use
Tapered
113.03 cm