AlphaProTech Critical Cover™ AlphaGuard™ Lab Coats
Soft and breathable multilayer SMS material provides comfort in critical environments. Choose from elastic or knit wrist styles in blue or white colors.
$141.68 - $222.89
Specifications
Product Type | Controlled Environments Lab Coat |
---|---|
Material | SMS |
Disposable | Single-use |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Price | Quantity | ||||||
19-130-1466A
|
AlphaProTech
LC326321 |
Case of 30 Each for $168.17
|
|
||||||
19-130-1466E
|
AlphaProTech
LC326325 |
Case of 30 Each for $209.22
|
|
||||||
19-130-1466D
|
AlphaProTech
LC326324 |
Case of 30 Each for $168.17
|
|
||||||
19-130-1466C
|
AlphaProTech
LC326323 |
Case of 30 Each for $141.68
|
|
||||||
19-130-1466B
|
AlphaProTech
LC326322 |
Case of 30 Each for $168.17
|
|
||||||
19-130-1466G
|
AlphaProTech
LC326329 |
Case of 30 Each for $222.89
|
|
||||||
19-130-1466F
|
AlphaProTech
LC326326 |
Case of 30 Each for $202.92
|
|
||||||
19-130-1467C
|
AlphaProTech
LC326223 |
Case of 30 Each for $186.04
|
|
||||||
19-130-1467B
|
AlphaProTech
LC326222 |
Case of 30 Each for $186.04
|
|
||||||
19-130-1467E
|
AlphaProTech
LC326225 |
Case of 30 Each for $194.74
|
|
||||||
19-130-1467D
|
AlphaProTech
LC326224 |
Case of 30 Each for $186.04
|
|
||||||
19-130-1467F
|
AlphaProTech
LC326226 |
Case of 30 Each for $204.46
|
|
||||||
19-130-4252
|
AlphaProTech
LC316321 |
Case of 30 Each for $168.17
|
|
||||||
19-130-4253
|
AlphaProTech
LC316322 |
Case of 30 Each for $168.17
|
|
||||||
19-130-4255
|
AlphaProTech
LC316324 |
Case of 30 Each for $165.00
|
|
||||||
19-130-4256
|
AlphaProTech
LC316325 |
Case of 30 Each for $164.17
|
|
||||||
19-130-4254
|
AlphaProTech
LC316323 |
Case of 30 Each for $155.71
|
|
||||||
19-130-4257
|
AlphaProTech
LC316326 |
Case of 30 Each for $171.97
|
|
||||||
Description
Soft and breathable multilayer SMS material provides comfort in critical environments. Choose from elastic or knit wrist styles in blue or white colors.
- Soft, breathable and fluid repellent
- Static dissipative
- Ultra clean, low particle count
Specifications
Controlled Environments Lab Coat | |
SMS | |
Single-use | |
Non-sterile |