AlphaProTech Critical Cover™ AquaTrak™ Ankle High Boot Covers
Boot covers provide anti-skid properties in wet and dry conditions. Two styles to choose from include seamless sonic welded or stress-resistant serged seams.
Supplier:  AlphaProTech BT91413B
 View more versions of this product
Catalog No. 19-130-4192
Explore available promotions
Description
Description
  • Low particle counts
  • Available as seamless sole with sonic welded seams or elastic all around with serged seams
Specifications
Specifications
X-Large
Blue
Non-sterile
Anti-skid
Black
16 in.
18 in.
Fluid, Dry Particulates
AquaTrak™
Single-use
Elastic Top with Ankle Ties
Sonic Welded
100 Pairs/Cs.
40.64 cm
45.72 cm
Controlled Environments Boot Covers