missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AlphaProTech Critical Cover™ AquaTrak™ UltraGrip™ Assurance™ Boot Covers

The combination of the UltraGrip sole with our AquaTrak upper material offers the ultimate in safety

$552.38 - $735.96

Specifications

Material AquaTrak™
Color White
Closure Type Elastic Top and Ankles
Seam Type Serged
For Use With (Application) Ideal for industrial use
View More Specs

Products 3
Catalog Number Mfr. No. Size Thread Color Price Quantity  
Catalog Number Mfr. No. Size Thread Color Price Quantity  
19-125-025
View Documents
AlphaProTech
BTAT1812B
Universal White
Case of 100 Pair for $674.32
 
19-125-026
View Documents
AlphaProTech
BTAT1813B
X-Large Black
Case of 100 Pair for $735.96
 
19-125-027
View Documents
AlphaProTech
BTAT1815B
Medium Yellow
Case of 100 Pair for $552.38
 
Description

Description

  • Durable, clean and impervious to fluids, particulates and several chemicals
  • Non-skid UltraGrip sole exhibits outstanding traction on a wide range of smooth to highly textured surfaces and works well on many surfaces contaminated with silicone
  • Sterility Assurance Level: Method 1 validated and terminally sterilized using gamma irradiation to yield a minimum SAL of 10-4
  • Processed to ensure 1 or fewer colony forming units in 10,000 garments
  • Non-conductive, serged seams, individually bagged in pairs

For industrial use only.

Specifications

Specifications

AquaTrak™
Elastic Top and Ankles
Ideal for industrial use
White
Serged
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'