AlphaProTech Critical Cover™ SureGrip™ Shoe Covers
Shoe covers with serged seams are constructed from a durable material with inherent anti-skid properties for dry environments. Also available in conductive styles for ESD concerns.
Supplier:  AlphaProTech SHX1223B
 View more versions of this product
Catalog No. 19-523-02
Explore available promotions
Description
Description
  • Constructed of SureGrip proprietary material
  • Inherently nonskid bottom
  • Elastic cuff with serged seams
  • Thread colors provide size identification
  • ESD styles and gaylord packaging (30 individual bags of 300 each per master carton) available
Specifications
Specifications
X-Large
Blue
Non-sterile
Non-skid
Black
Standard
43.18 cm
16.51 cm
SureGrip™
Single-use
Elastic Ankle
Serged
150 Pairs/Cs.
17 in.
6.5 in.
Controlled Environments Shoe Covers