AlphaProTech Critical Cover™ SureGrip™ Shoe Covers
Shoe covers with serged seams are constructed from a durable material with inherent anti-skid properties for dry environments. Also available in conductive styles for ESD concerns.
$152.22 - $4894.40
Specifications
Material SureGrip™
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
View More Specs
Products 5
Catalog Number Mfr. No. Size Thread Color Packaging Type Price Quantity  
Catalog Number Mfr. No. Size Thread Color Packaging Type Price Quantity  
19-520-31
View Documents
AlphaProTech
SHX1222B
Universal White Standard
Case of 300 Each for $152.22
19-023-020
View Documents
AlphaProTech
SHX1224B
2X-Large Blue Standard
Case of 300 Each for $168.99
19-065-337A
View Documents
AlphaProTech
SHX1273B
X-Large Black Gaylord Packaging
Case of 4500 Pair for $4,607.26
19-065-337B
View Documents
AlphaProTech
SHX1274B
2X-Large Blue Gaylord Packaging
Case of 9000 Each for $4,894.40
19-523-02
View Documents
AlphaProTech
SHX1223B
X-Large Black Standard
Case of 300 Each for $191.34
Description
Description
  • Constructed of SureGrip proprietary material
  • Inherently nonskid bottom
  • Elastic cuff with serged seams
  • Thread colors provide size identification
  • ESD styles and gaylord packaging (30 individual bags of 300 each per master carton) available
Specifications
Specifications
SureGrip™
Single-use
Elastic Ankle
Serged
Blue
Non-sterile
Non-skid
Controlled Environments Shoe Covers
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'