missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell™ AlphaTec™ Chemical Protective Apron with Sleeves
Ansell™ AlphaTec™ Chemical Protective Apron with Sleeves for exceptional chemical protection
Supplier: Ansell™ GR40T9221507
Description
- Velcro fastening to neck
- Tie fastening at the waist
- Double cuff with knitted inner cuff
- Welded and taped seams
- Applications:
- Contaminated environments
- Chemical handling
- Filling, blending and charging of raw materials
- Tranferring liquids and solids
- Opening and draining pumps valves or lines
- Loading and unloading of process equipment
- Raw material sample collection
- Concentration mixing and palletisation
- Hazmat response
- Assembly and disassembly of large pipes
Specifications
Protective Apron | |
Multi-layer Barrier Laminate Fabric | |
Hook and Loop | |
Double Inner Knit | |
103 gsm |
3X-Large | |
Green | |
Non-sterile | |
CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126) | |
Ultrasonically Welded and Taped |