missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ansell™ AlphaTec™ Chemical Protective Apron with Sleeves

Ansell™ AlphaTec™ Chemical Protective Apron with Sleeves for exceptional chemical protection

Supplier:  Ansell™ GR40T9221509

 View more versions of this product

Catalog No. 19-010-378


Explore available promotions

Description

Description

  • Velcro fastening to neck
  • Tie fastening at the waist
  • Double cuff with knitted inner cuff
  • Welded and taped seams
  • Applications:
    • Contaminated environments
    • Chemical handling
    • Filling, blending and charging of raw materials
    • Tranferring liquids and solids
    • Opening and draining pumps valves or lines
    • Loading and unloading of process equipment
    • Raw material sample collection
    • Concentration mixing and palletisation
    • Hazmat response
    • Assembly and disassembly of large pipes
Specifications

Specifications

Protective Apron
Multi-layer Barrier Laminate Fabric
Hook and Loop
Double Inner Knit
103 gsm
5X-Large
Green
Non-sterile
CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126)
Ultrasonically Welded and Taped
Videos
SDS
Documents

Documents

Promotions

Promotions