missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell AlphaTec™ Solvex™ 37-175 Nitrile Gloves With Cotton Flocking
Industrial 15 mil thick nitrile gloves offer versatile chemical protection across a diverse range of applications.
Supplier: Ansell 3717511100
Description
Industrial 15 mil thick nitrile gloves offer versatile chemical protection across a diverse range of applications.
- Nitrile compound provides combination of chemical resistance and strength
- Cotton flocking, reducing sweat for the wearer
- Sandpatch finish giving the glove a smooth surface
- Fully reusable, with an unequaled abrasion resistance
- Complies with FDA food handling requirements (21 CFR 177.2600)
Specifications
ANSI 4 | |
CE 0086, EN 420:2003 + A1:2009, Category III | |
Green | |
Nitrile | |
15 mil | |
EN 388:4101X, EN 388:4101 | |
Type A | |
Bacteria, Fungi WITH Virus Protection | |
Gauntlet | |
Cotton Flock | |
37-175 10 117276 | |
13 in. |
FDA 21 CFR 177.2600, ASTM F1671 | |
Yes | |
Yes | |
10 | |
0.38 mm | |
No | |
JKLOPT | |
Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work | |
Sandpatch | |
Lined | |
AlphaTec™ Solvex™ 37-175 | |
33 cm |