missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell AlphaTec™ Solvex™ 37-175 Nitrile Gloves With Cotton Flocking
Industrial 15 mil thick nitrile gloves offer versatile chemical protection across a diverse range of applications.
$.67 - $2.42
Specifications
ANSI Abrasion Rating | ANSI 4 |
---|---|
Certifications/Compliance | FDA 21 CFR 177.2600, ASTM F1671 |
Certifications/Compliance (Europe) | CE 0086, EN 420:2003 + A1:2009, Category III |
Chlorinated | Yes |
Color | Green |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
11-391-139A
|
Ansell
3717511060 |
6 |
Pair for $0.67
Case of 144 Pair for $96.48
|
|
|||||
11-391-139B
|
Ansell
3717511070 |
7 |
Pair for $2.09
Case of 144 Pair for $250.56
|
|
|||||
11-391-139C
|
Ansell
3717511080 |
8 |
Pair for $2.42
Case of 144 Pair for $289.44
|
|
|||||
11-391-139D
|
Ansell
3717511090 |
9 |
Pair for $2.42
Case of 144 Pair for $289.44
|
|
|||||
11-391-139E
|
Ansell
3717511100 |
10 |
Pair for $2.42
Case of 144 Pair for $289.44
|
|
|||||
11-391-139F
|
Ansell
3717511110 |
11 |
Pair for $2.09
Case of 144 Pair for $250.56
|
|
|||||
Description
Industrial 15 mil thick nitrile gloves offer versatile chemical protection across a diverse range of applications.
- Nitrile compound provides combination of chemical resistance and strength
- Cotton flocking, reducing sweat for the wearer
- Sandpatch finish giving the glove a smooth surface
- Fully reusable, with an unequaled abrasion resistance
- Complies with FDA food handling requirements (21 CFR 177.2600)
Specifications
ANSI 4 | |
CE 0086, EN 420:2003 + A1:2009, Category III | |
Green | |
Nitrile | |
0.38 mm | |
No | |
JKLOPT | |
Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work | |
Sandpatch | |
Lined | |
13 in. |
FDA 21 CFR 177.2600, ASTM F1671 | |
Yes | |
Yes | |
15 mil | |
EN 388:4101X, EN 388:4101 | |
Type A | |
Bacteria, Fungi WITH Virus Protection | |
Gauntlet | |
Cotton Flock | |
AlphaTec™ Solvex™ 37-175 | |
33 cm |