missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alumina R, MP EcoChrom™, MP Biomedicals™
MP EcoChrom™ Alumin R is an acid alumina, developed for use in isotope chemistry.
Supplier: MP Biomedicals Inc 0206034
Description
- MP EcoChrom™ Alumina R catalyses a variety of reactions that are useful industrially.
- It is widely used to remove water from gas streams.
- Also used for its hardness and strength.
- It is widely used as an abrasive, including as a much less expensive substitute for industrial diamond.
- Many types of sandpaper use aluminium oxide crystals.
Specifications
Alumina R | |
Pink to White | |
4.3 ± 0.1 | |
500 g | |
MFCD00003424 | |
Insoluble in water. | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
3.94 g/cm3 (at 20°C) |
ca. 2072 °C | |
1344-28-1 | |
Free-flowing Powder | |
Al2O3 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminium(3+) trioxidandiide | |
9989226 | |
At 20 °C (not applicable) |
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
dialuminium(3+) trioxidandiide |
Safety and Handling
EINECSNumber : 215-691-6
RTECSNumber : BD1200000
Recommended Storage : Store at room temperature.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.