Learn More
Aluminum isopropoxide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 205170010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Aluminum isopropoxide | |
129.0°C to 136.0°C | |
125.0°C to 130.0°C (38.0 mmHg) | |
98% | |
C9H21AlO3 | |
MFCD00008870 | |
01,35; 03,10; 04,15; 05,14; 06,19; 08,15; 09,14; 11,29; 13,265; 15,16 | |
aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
aluminum;propan-2-olate | |
11143 | |
98% |
555-31-7 | |
Yellow | |
Authentic | |
Plastic bottle | |
[(CH3)2CHO]3Al | |
1 kg | |
15, 342 | |
Solubility in water: reacts. Other solubilities: freely soluble in ethanol | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] | |
204.25 | |
204.25 | |
Crystalline Powder |
Chemical Identifiers
555-31-7 | |
204.25 | |
SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
11143 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] |
C9H21AlO3 | |
MFCD00008870 | |
aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
aluminum;propan-2-olate |
Safety and Handling
GHS H Statement
Flammable solid.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing d
GHS Signal Word: Danger
EINECSNumber : 209-090-8
RUO – Research Use Only