Learn More
Aluminum isopropoxide, 98%, Thermo Scientific Chemicals
$84.75 - $124.55
Chemical Identifiers
CAS | 555-31-7 |
---|---|
Molecular Formula | C9H21AlO3 |
Molecular Weight (g/mol) | 204.25 |
MDL Number | MFCD00008870 |
InChI Key | SMZOGRDCAXLAAR-UHFFFAOYSA-N |
Synonym | aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate |
PubChem CID | 11143 |
IUPAC Name | aluminum;propan-2-olate |
SMILES | CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC205175000
|
Thermo Scientific Chemicals
205175000 |
500 g | Plastic bottle |
Each for $84.75
|
|
||||
AC205170010
|
Thermo Scientific Chemicals
205170010 |
1 kg | Plastic bottle |
Each for $124.55
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
555-31-7 | |
204.25 | |
SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
11143 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] |
C9H21AlO3 | |
MFCD00008870 | |
aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
aluminum;propan-2-olate |
Specifications
555-31-7 | |
Yellow | |
Authentic | |
Plastic bottle | |
[(CH3)2CHO]3Al | |
500 g | |
15, 342 | |
Solubility in water: reacts. Other solubilities: freely soluble in ethanol | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] | |
204.25 | |
204.25 | |
Crystalline Powder |
129.0°C to 136.0°C | |
125.0°C to 130.0°C (38.0 mmHg) | |
98% | |
C9H21AlO3 | |
MFCD00008870 | |
01,35; 03,10; 04,15; 05,14; 06,19; 08,15; 09,14; 11,29; 13,265; 15,16 | |
aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
aluminum;propan-2-olate | |
11143 | |
98% | |
Aluminum isopropoxide |
Safety and Handling
GHS H Statement
Flammable solid.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing d
GHS Signal Word: Danger
EINECSNumber : 209-090-8
RUO – Research Use Only