missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum isopropoxide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 205175000

 View more versions of this product

Catalog No. AC205175000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Aluminum isopropoxide
129.0°C to 136.0°C
125.0°C to 130.0°C (38.0 mmHg)
98%
C9H21AlO3
MFCD00008870
01,35; 03,10; 04,15; 05,14; 06,19; 08,15; 09,14; 11,29; 13,265; 15,16
aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate
SMZOGRDCAXLAAR-UHFFFAOYSA-N
aluminum;propan-2-olate
11143
98%
555-31-7
Yellow
Authentic
Plastic bottle
[(CH3)2CHO]3Al
500 g
15, 342
Solubility in water: reacts. Other solubilities: freely soluble in ethanol
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3]
204.25
204.25
Crystalline Powder

Chemical Identifiers

555-31-7
204.25
SMZOGRDCAXLAAR-UHFFFAOYSA-N
11143
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3]
C9H21AlO3
MFCD00008870
aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate
aluminum;propan-2-olate
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing d

GHS Signal Word: Danger

EINECSNumber : 209-090-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only