Learn More
Aluminum sulfate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 192430250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
Aluminum sulfate | |
Extra Pure | |
White | |
25 g | |
Al2O12S3 | |
MFCD00003423 | |
aluminum sulfate, aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2 | |
DIZPMCHEQGEION-UHFFFAOYSA-H | |
dialuminum;trisulfate | |
24850 | |
342.13 | |
Trace Metal Basis |
770°C | |
10043-01-3 | |
Chunks and/or Powder | |
15% min. (Al) Typical (Complexometry) | |
Al2(SO4)3 | |
15, 360 | |
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol | |
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] | |
342.13 | |
CHEBI:74768 | |
100.00% | |
Glass bottle |
Chemical Identifiers
10043-01-3 | |
342.13 | |
DIZPMCHEQGEION-UHFFFAOYSA-H | |
24850 | |
dialuminum;trisulfate |
Al2O12S3 | |
MFCD00003423 | |
aluminum sulfate, aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2 | |
CHEBI:74768 | |
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 233-135-
RUO – Research Use Only