Learn More
Aluminum sulfate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals
$127.73 - $850.21
Chemical Identifiers
CAS | 10043-01-3 |
---|---|
Molecular Formula | Al2O12S3 |
Molecular Weight (g/mol) | 342.13 |
MDL Number | MFCD00003423 |
InChI Key | DIZPMCHEQGEION-UHFFFAOYSA-H |
Synonym | aluminum sulfate, aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2 |
PubChem CID | 24850 |
ChEBI | CHEBI:74768 |
IUPAC Name | dialuminum;trisulfate |
SMILES | [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC192430050
|
Thermo Scientific Chemicals
192430050 |
5 g | Glass bottle |
Each for $127.73
|
|
||||
AC192430250
|
Thermo Scientific Chemicals
192430250 |
25 g | Glass bottle |
Each for $302.07
|
|
||||
AC192431000
|
Thermo Scientific Chemicals
192431000 |
100 g | Glass bottle |
Each for $850.21
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
10043-01-3 | |
342.13 | |
DIZPMCHEQGEION-UHFFFAOYSA-H | |
24850 | |
dialuminum;trisulfate |
Al2O12S3 | |
MFCD00003423 | |
aluminum sulfate, aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2 | |
CHEBI:74768 | |
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] |
Specifications
770°C | |
White | |
5 g | |
Al2O12S3 | |
MFCD00003423 | |
aluminum sulfate, aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2 | |
DIZPMCHEQGEION-UHFFFAOYSA-H | |
dialuminum;trisulfate | |
24850 | |
342.13 | |
Trace Metal Basis | |
Aluminum sulfate, Extra Pure |
10043-01-3 | |
Chunks and/or Powder | |
15% min. (Al) Typical (Complexometry) | |
Al2(SO4)3 | |
15, 360 | |
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol | |
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] | |
342.13 | |
CHEBI:74768 | |
100.00% | |
Glass bottle |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 233-135-
RUO – Research Use Only