missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRC-120(H), ion exchange resin

CAS: 78922-04-0 | C13H10ClNO4S | 311.736 g/mol

$170.71 - $170.71

Chemical Identifiers

CAS 78922-04-0
Molecular Formula C13H10ClNO4S
Molecular Weight (g/mol) 311.736
MDL Number MFCD00132707
InChI Key APBOVLPLJFJSRI-UHFFFAOYSA-N
Synonym 3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid
PubChem CID 8190984
IUPAC Name 3-[(3-chlorophenyl)sulfonylamino]benzoic acid
SMILES C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL142850B
View Documents
Thermo Scientific Chemicals
L142850B
1000 g
Each for $170.71
Only null left
 
Description

Description

Amberlite™ IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite|r IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

78922-04-0
311.736
APBOVLPLJFJSRI-UHFFFAOYSA-N
8190984
C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O
C13H10ClNO4S
MFCD00132707
3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid
3-[(3-chlorophenyl)sulfonylamino]benzoic acid

Specifications

78922-04-0
MFCD00132707
14,382
3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid
APBOVLPLJFJSRI-UHFFFAOYSA-N
3-[(3-chlorophenyl)sulfonylamino]benzoic acid
8190984
C13H10ClNO4S
1.28
1000 g
Insoluble in water.
C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O
311.736
Amberlite™ IR-120(H), ion exchange resin
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P280-P305+P351+P338-P310

H318

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only