Learn More
Invitrogen™ Recombinant Proteinase K Solution (20 mg/mL)
Recombinant Proteinase K is a nonspecific serine protease that is useful for general digestion of proteins.
$119.00 - $377.00
Specifications
Concentration | 20 mg/mL |
---|---|
Host Species | T. album |
Content And Storage | Store at -20°C. Shipped at ambient temperature. |
Shipping Condition | Dry Ice |
Product Type | Proteinase K |
Description
Recombinant Proteinase K is a nonspecific serine protease that is useful for general digestion of proteins.
Proteinase K remains active:
• Over a wide pH range (optimal activity between 6.5 and 9.5)
• Under denaturing conditions (e.g., in the presence of SDS or urea)
• In the presence of metal chelating agents (e.g., EDTA)
• At comparatively high temperatures (optimum digestion temperature is 65°C)
Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.
Performance and quality testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.
Unit definition
One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.
Order Info
Shipping Condition: Dry Ice
Specifications
20 mg/mL | |
Store at -20°C. Shipped at ambient temperature. |
|
Proteinase K | |
Liquid |
T. album | |
Dry Ice | |
Ambion™ |
For Research Use Only. Not for use in diagnostic procedures.