missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Ambroxide 98.0+%, TCI America™

Supplier:  TCI America A247225G

 View more versions of this product

Catalog No. A247225G



Description

Description

Specifications

Specifications

(-)-Ambroxide
6790-58-5
25 g
C16H28O
1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane
CC12CCC3C(C)(C)CCCC3(C)C1CCO2
236.40
CHEBI:78307
≥98.0% (GC)
77°C
White
Crystal-Powder at 20°C
MFCD00134491
YPZUZOLGGMJZJO-UHFFFAOYNA-N
3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan
10857465
236.40

Chemical Identifiers

6790-58-5
236.40
YPZUZOLGGMJZJO-UHFFFAOYNA-N
10857465
3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan
C16H28O
MFCD00134491
1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane
CHEBI:78307
CC12CCC3C(C)(C)CCCC3(C)C1CCO2
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions