missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(-)-Ambroxide 98.0+%, TCI America™
$94.24 - $321.40
Chemical Identifiers
CAS | 6790-58-5 |
---|---|
Molecular Formula | C16H28O |
Molecular Weight (g/mol) | 236.40 |
MDL Number | MFCD00134491 |
InChI Key | YPZUZOLGGMJZJO-UHFFFAOYNA-N |
Synonym | 1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane |
PubChem CID | 10857465 |
ChEBI | CHEBI:78307 |
IUPAC Name | 3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan |
SMILES | CC12CCC3C(C)(C)CCCC3(C)C1CCO2 |
Chemical Identifiers
6790-58-5 | |
236.40 | |
YPZUZOLGGMJZJO-UHFFFAOYNA-N | |
10857465 | |
3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan |
C16H28O | |
MFCD00134491 | |
1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane | |
CHEBI:78307 | |
CC12CCC3C(C)(C)CCCC3(C)C1CCO2 |
Specifications
77°C | |
White | |
Crystal-Powder at 20°C | |
MFCD00134491 | |
YPZUZOLGGMJZJO-UHFFFAOYNA-N | |
3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan | |
10857465 | |
236.40 | |
(-)-Ambroxide |
6790-58-5 | |
5 g | |
C16H28O | |
1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane | |
CC12CCC3C(C)(C)CCCC3(C)C1CCO2 | |
236.40 | |
CHEBI:78307 | |
≥98.0% (GC) |
Safety and Handling
TSCA : Yes