missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Amersham™ ECL™ Prime Western Blotting Detection Reagent
Enables the use of highly diluted primary and secondary antibodies without reducing sensitivity. Cytiva Lifescience™ Amersham™ ECL™ Prime Western Blotting Detection Reagent produces high signal intensity and sensitivity.
$479.39 - $917.85
Specifications
Product Type | ECL Western Blot Detection Reagent |
---|---|
For Use With (Application) | Sensitive Quantitative or Confirmatory Western Blot |
Sensitivity | ∼1 pg |
Content And Storage | 2°C to 8°C |
Includes: Solution A (luminol solution) and Solution B (peroxide solution)
Catalog Number | Mfr. No. | Kit Contents | Sufficient For | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Kit Contents | Sufficient For | Price | Quantity | ||||
45-002-401
|
Cytiva
RPN2232 |
Solution A (Luminol solution, 50mL) and Solution B (Peroxide solution, 50mL) | 1000cm2 membrane |
Each for $479.39
|
|
||||
45-010-090
|
Cytiva
RPN2236 |
Solution A (Luminol solution, 3 × 50mL) and Solution B (Peroxide solution, 3 × 50mL) | 3000cm2 membrane |
Each for $917.85
|
|
||||
Description
- Stable signal enables multiple exposures and makes reagent suitable for large experimental series
- Provides convenient handling time between detection and completion of experiment
- Optimized for imaging with ImageQuant™ LAS 4000 (CCD-based imaging), Amersham Hybond™ P (PVDF) membranes and compatible with Amersham Hyperfilm™, Amersham Hybond- ECL (nitrocellulose) membranes
- Also compatible with Rainbow™ Molecular Weight Markers and Amersham ECL DualVue™ Western Blotting Markers
Education and research
Specifications
ECL Western Blot Detection Reagent | |
Sensitive Quantitative or Confirmatory Western Blot | |
∼1 pg | |
2°C to 8°C |