missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Acetate, ∽99%, (Crystalline), MP Biomedicals

Supplier:  MP Biomedicals Inc 0210065102

 View more versions of this product

Catalog No. ICN10065102


Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Acetate
Crystalline
∽99%
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CC(=O)[O-].[NH4+]
77.083
CHEBI:62947
631-61-8
2 kg
C2H7NO2
USFZMSVCRYTOJT-UHFFFAOYSA-N
azanium;acetate
517165
77.1

Chemical Identifiers

631-61-8
77.083
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CHEBI:62947
CC(=O)[O-].[NH4+]
C2H7NO2
USFZMSVCRYTOJT-UHFFFAOYSA-N
517165
azanium;acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS Documents

Documents

Promotions

Promotions