missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, for analysis

CAS: 506-87-6 | CH8N2O3 | 96.09 g/mol

$96.90 - $556.95

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.09
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC196651000
SDS View Documents
Thermo Scientific Chemicals
196651000
100 g Plastic bottle
Each for $96.90
Only null left
 
AC196655000
SDS View Documents
Thermo Scientific Chemicals
196655000
500 g Plastic bottle
Each for $149.11
Only null left
 
AC196650025
SDS View Documents
Thermo Scientific Chemicals
196650025
2.5 kg Plastic bottle
Each for $556.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

506-87-6
96.09
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
517111
C(=O)([O-])[O-].[NH4+].[NH4+]
CH8N2O3
MFCD00010890
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
diazanium;carbonate

Specifications

58.0°C
White
100 g
(NH4)2CO3
15, 505
Solubility in water: soluble (320g/L at 20°C)
C(=O)([O-])[O-].[NH4+].[NH4+]
96.09
96.09
Analytical
Ammonium carbonate, p.a.
506-87-6
Crystalline Powder
CH8N2O3
MFCD00010890
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
diazanium;carbonate
517111
≥30.0% (NH3)
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-058-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only