missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Hydroxide, 6.00 N, Ricca Chemical
Supplier: Ricca Chemical Company 64516
Specifications
Ammonium Hydroxide, 6.00N, Ricca Chemical | |
71.0,26.6 | |
H5NO | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 | |
Natural Poly Bottle |
1336-21-6,7732-18-5 | |
74.0,27.69 | |
MFCD00066650 | |
500 mL | |
N.O | |
35.05 | |
CHEBI:18219 |
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
amine hydrate |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
CHEBI:18219 | |
N.O |