missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Hydroxide, 6.00 N, Ricca Chemical
$125.14 - $263.00
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
IUPAC Name | amine hydrate |
SMILES | N.O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
645-16
|
Ricca Chemical Company
64516 |
500 mL | Natural Poly Bottle |
Each for $125.14
|
|
||||
645-32
|
Ricca Chemical Company
64532 |
1 L | Natural Poly Bottle |
Each for $263.00
|
|
||||
R06450004A
|
Ricca Chemical Company
R06450004A |
4 L | Natural Poly Bottle |
Each for $258.00
|
|
||||
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
amine hydrate |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
CHEBI:18219 | |
N.O |
Specifications
1336-21-6,7732-18-5 | |
74.0,27.69 | |
MFCD00066650 | |
500 mL | |
N.O | |
35.05 | |
CHEBI:18219 | |
Ammonium Hydroxide, 6.00N, Ricca Chemical |
71.0,26.6 | |
H5NO | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 | |
Natural Poly Bottle |