missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Hydroxide, 6.00 N, Ricca Chemical

Supplier:  Ricca Chemical Company R06450004A

 View more versions of this product

Catalog No. R06450004A


Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Hydroxide, 6.00N, Ricca Chemical
71.0,26.6
H5NO
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
Natural Poly Bottle
1336-21-6,7732-18-5
74.0,27.69
MFCD00066650
4 L
N.O
35.05
CHEBI:18219

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
amine hydrate
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
N.O
Videos
SDS
Documents

Documents

Promotions

Promotions