missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium hydroxide, ACS, 28.0-30.0% NH{3}, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 033285A1

 View more versions of this product

Catalog No. AA33285A1

Explore available promotions

Description

Description

Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silve, and platinum jewelry. It is an active component of Tollens' reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium hydroxide
1336-21-6
H5NO
0.90
Miscible with water.
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
35.05
Liquid
Colorless
28.0-30.0% NH3
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
1 kg
N.O
35.05
CHEBI:18219
ACS

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
N.O
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 215-647-6

RTECSNumber : BQ9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only