missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium hydroxide, ACS, 28.0-30.0% NH{3}, Thermo Scientific Chemicals

$49.48 - $302.18

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3328530
View Documents
Thermo Scientific Chemicals
03328530
250 g
Each for $49.48
 
AA33285A1
View Documents
Thermo Scientific Chemicals
033285A1
1 kg
Each for $86.47
 
AA33285D9
View Documents
Thermo Scientific Chemicals
033285D9
5 x 1 kg
Case of 5 Each for $302.18
 
Description

Description

Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silve, and platinum jewelry. It is an active component of Tollens' reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
N.O
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219

Specifications

Colorless
28.0-30.0% NH3
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
250 g
N.O
35.05
CHEBI:18219
ACS
Liquid
1336-21-6
H5NO
0.90 g/cm3
Miscible with water.
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
35.05
Ammonia-like
Ammonium hydroxide
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 215-647-6

RTECSNumber : BQ9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only