missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Phosphate Dibasic, Crystal, ACS, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A1240125GM

 View more versions of this product

Catalog No. 18608756


Explore available promotions

Description

Description

Specifications

Specifications

7783-28-0
7.7 to 8.1
H9N2O4P
MNNHAPBLZZVQHP-UHFFFAOYSA-N
phosphoric acid diamine
98%
Poly Bottle
1
125 g
MFCD00010891
N.N.OP(O)(O)=O
132.06
ACS

Chemical Identifiers

7783-28-0
132.06
MNNHAPBLZZVQHP-UHFFFAOYSA-N
N.N.OP(O)(O)=O
H9N2O4P
MFCD00010891
phosphoric acid diamine
Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.