missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate Dibasic, Crystal, ACS, 98%, Spectrum™ Chemical
$150.65 - $8180.53
Chemical Identifiers
| CAS | 7783-28-0 |
|---|---|
| Molecular Formula | H9N2O4P |
| Molecular Weight (g/mol) | 132.06 |
| MDL Number | MFCD00010891 |
| InChI Key | MNNHAPBLZZVQHP-UHFFFAOYSA-N |
| IUPAC Name | phosphoric acid diamine |
| SMILES | N.N.OP(O)(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18608755
|
Spectrum Chemical Mfg Cor
A124025GM |
25 g | Poly Bottle |
Each for $150.65
|
|
||||
|
18608756
|
Spectrum Chemical Mfg Cor
A1240125GM |
125 g | Poly Bottle |
Each for $291.76
|
|
||||
|
18608757
|
Spectrum Chemical Mfg Cor
A1240500GM |
500 g | Poly Bottle |
Each for $502.62
|
|
||||
|
18608758
|
Spectrum Chemical Mfg Cor
A124025KG |
2.5 kg | Poly Bottle |
Each for $1,982.21
|
|
||||
|
18608759
|
Spectrum Chemical Mfg Cor
A124012KG |
12 kg | Poly Pail |
Each for $4,024.59
|
|
||||
|
18608760
|
Spectrum Chemical Mfg Cor
A124050KGBL |
50 kg | Fiber Drum |
Each for $8,180.53
|
|
||||
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.