missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aniline sulfate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1556322

 View more versions of this product

Catalog No. AAA1556322


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Aniline sulfate
1.377
(C6H5NH2)2·H2SO4
100 g
3729533
Soluble in water
98%
542-16-5
Odorless
MFCD00013110
UN2811
Benzenamine sulfate 2:1
284.34

Chemical Identifiers

542-16-5
(C6H5NH2)2·H2SO4
MFCD00013110
Benzenamine sulfate 2:1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H372-H341-H351-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of causing cancer.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P310-P312-P330-P333+P313-P361-P363-P501c

H301+H311+H331-H317-H318-H341-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 208-805-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only