missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aniline sulfate, 97%, Thermo Scientific Chemicals

$80.36 - $257.49

Chemical Identifiers

CAS 542-16-5
Molecular Formula (C6H5NH2)2·H2SO4
MDL Number MFCD00013110
Synonym Benzenamine sulfate 2:1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1556322
View Documents
Thermo Scientific Chemicals
A1556322
100 g
Each for $80.36
 
AAA1556336
View Documents
Thermo Scientific Chemicals
A1556336
500 g
Each for $257.49
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

542-16-5
(C6H5NH2)2·H2SO4
MFCD00013110
Benzenamine sulfate 2:1

Specifications

542-16-5
Odorless
MFCD00013110
UN2811
Benzenamine sulfate 2:1
284.34
Aniline sulfate
1.377
(C6H5NH2)2·H2SO4
100 g
3729533
Soluble in water
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H372-H341-H351-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of causing cancer.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P310-P312-P330-P333+P313-P361-P363-P501c

H301+H311+H331-H317-H318-H341-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 208-805-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only