Fisherbrand™ Animal Ear Punch
These instruments are available in multiple designs, including thumb-style, plier-style, or scissor-style punches. 
Supplier:  Fisherbrand™ 10401164
 View more versions of this product
Catalog No. 13-820-063
Explore available promotions
Description
Description
Fisherbrand™ Animal Ear Punches are made from corrosion-resistant ASTM (899-94) Series 400 stainless steel. These instruments are available in multiple designs, including thumb-style, plier-style, or scissor-style punches. Both the plier-style and scissor-style punches feature a longer handle for improved grip and handling.
  • Surgical grade stainless steel
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Certifications
ISO 9001-02, GMP, cGMP
Specifications
Specifications
Autoclavable
Silver
Animal Ear
Premium
Gamma/ETO/Autoclave
4.25 in.
0.04 in.
ISO, cGMP
Surgical grade stainless steel
Lab Punch
Stainless Steel
Plier-Style
10.8 cm
1 mm