missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Anisoin, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2155936

 View more versions of this product

Catalog No. AAB2155936


Explore available promotions

Description

Description

Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.

Solubility
Soluble in water, ethanol and acetone.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Specifications

Specifications

Anisoin
110°C to 114°C
MFCD00008411
3211892
Soluble in water,ethanol and acetone.
COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1
272.30
272.3
119-52-8
C16H16O4
500 g
anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one
LRRQSCPPOIUNGX-UHFFFAOYNA-N
2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone
95415
97%

Chemical Identifiers

119-52-8
272.30
LRRQSCPPOIUNGX-UHFFFAOYNA-N
95415
COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1
C16H16O4
MFCD00008411
anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one
2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-330-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only