Learn More
Anisoin, 97%, Thermo Scientific Chemicals
$95.33 - $878.40
Chemical Identifiers
CAS | 119-52-8 |
---|---|
Molecular Formula | C16H16O4 |
Molecular Weight (g/mol) | 272.30 |
MDL Number | MFCD00008411 |
InChI Key | LRRQSCPPOIUNGX-UHFFFAOYNA-N |
Synonym | anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one |
PubChem CID | 95415 |
IUPAC Name | 2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone |
SMILES | COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2155914
|
Thermo Scientific Chemicals
B2155914 |
25 g |
Each for $95.33
|
|
||||||
AAB2155936
|
Thermo Scientific Chemicals
B2155936 |
500 g |
Each for $878.40
|
|
||||||
Description
Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAnisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.
Solubility
Soluble in water, ethanol and acetone.
Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Chemical Identifiers
119-52-8 | |
272.30 | |
LRRQSCPPOIUNGX-UHFFFAOYNA-N | |
95415 | |
COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1 |
C16H16O4 | |
MFCD00008411 | |
anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one | |
2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone |
Specifications
119-52-8 | |
C16H16O4 | |
25 g | |
anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one | |
LRRQSCPPOIUNGX-UHFFFAOYNA-N | |
2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone | |
95415 | |
97% |
110°C to 114°C | |
MFCD00008411 | |
3211892 | |
Soluble in water,ethanol and acetone. | |
COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1 | |
272.30 | |
272.3 | |
Anisoin |
Safety and Handling
EINECSNumber : 204-330-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only