Ansell HyFlex™ 11-501 Aramid Fiber Stretch Armor Gloves, Nitrile Coated
Ergonomic design reduces hand fatigue and provides a fit that mimics bare hands. Nitrile foam coating enhances tactile sensitivity and grip capabilities in dry or light oil conditions.
Supplier: Ansell 1150111080
Description
- Intercept Technology™ raises the protection & dexterity standards for cut-resistant gloves
- Aramid fiber provides an additional level of protection
- Ergonomic design provides bare-hand closeness and reduces hand fatigue
- Nitrile foam coating enhances grip capabilities and tactile sensitivity
Specifications
ANSI 3 | |
Blue | |
Specific | |
Cut-resistant Gloves | |
Automotive Repairs, Machinery/Equipment Operation, Metal Fabrication, and Chemical Handling | |
11-501 |
Nitrile | |
ANSI A5 | |
Aramid Fiber | |
8 | |
Knit |