Ansell HyFlex™ 11-600B Black Nylon Gloves with Gray Polyurethane Coating
Polyurethane coating on the palm improves tactile sensitivity, while the dark color palette is useful for applications involving dirt or grease.
Supplier:  Ansell 1160011080
 View more versions of this product
Catalog No. 19-130-3866
Explore available promotions
Description
Description
  • Designed for light & delicate applications in dry environments
  • Bare-handed dexterity with protection for light duty applications
  • Polyurethane coating on the palm enhances tactile sensitivity
  • Dark color palette is ideal for applications involving dirt or grease
Specifications
Specifications
ANSI 3
Black
Specific
Seamless Knit Gloves
Automotive, Machinery/Equipment, and Metal Fabrication
11-600B
15
Polyurethane
ANSI A1
Nylon
8
Knit
11-600B