missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell HyFlex™ 11-600B Black Nylon Gloves with Gray Polyurethane Coating
Polyurethane coating on the palm improves tactile sensitivity, while the dark color palette is useful for applications involving dirt or grease.
$35.26 - $38.91
Specifications
ANSI Abrasion Rating | ANSI 3 |
---|---|
Coating Material | Polyurethane |
Cut Resistance Rating | ANSI A1 |
Hand | Specific |
Material | Nylon |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-130-3864
|
Ansell
1160011060 |
6 |
Dozen of 12 Pair for $38.91
Case of 12 Dozen for $446.04
|
|
|||||
19-130-3865
|
Ansell
1160011070 |
7 |
Dozen of 12 Pair for $38.91
Case of 12 Dozen for $446.04
|
|
|||||
19-130-3866
|
Ansell
1160011080 |
8 |
Dozen of 12 Pair for $38.91
Case of 12 Dozen for $106.56
|
|
|||||
19-130-3867
|
Ansell
1160011100 |
10 |
Dozen of 12 Pair for $35.26
Case of 12 Dozen for $404.16
|
|
|||||
Description
- Designed for light & delicate applications in dry environments
- Bare-handed dexterity with protection for light duty applications
- Polyurethane coating on the palm enhances tactile sensitivity
- Dark color palette is ideal for applications involving dirt or grease
Specifications
ANSI 3 | |
ANSI A1 | |
Nylon | |
Knit | |
11-600B |
Polyurethane | |
Specific | |
Automotive, Machinery/Equipment, and Metal Fabrication | |
11-600B | |
15 |