Ansell HyFlex™ 74-048 Lightweight Cut Protection Gloves
Lightweight gloves provide a high-level of cut protection and dexterity. Can be used as a liner under another glove.
Supplier:  Ansell 7404811090
 View more versions of this product
Catalog No. 19-160-390D
Explore available promotions
Description
Description
  • Knitted, uncoated construction with DSM Dyneema™ liner
  • Suitable for a variety of food handling and meat cutting applications
  • Extended, anti-shrink Tuff-Cuff™ cuffs for additional protection
  • For most knife-hand, and some off-hand, applications
  • Ideal for beef, pork, poultry, seafood, vegetable and fruit processing
Specifications
Specifications
ANSI 3
Knit
ANSI A6
Dyneema
Large
Food processing
Dyneema™
HyFlex™ 74-048
23.5 cm
EN 420:2003 + A1:2009
Gray
Yes
Cut-resistant Gloves
EN 388:4X2XF, EN388:454X
Extended
74-048 L 245314
9.3 in.