Ansell AlphaTec™ 56-600 Urethane Aprons
Urethane aprons designed for comfort and extended wear. Offers a combination of chemical protection, flexibility, and toughness
Supplier:  Ansell YU48
 View more versions of this product
Catalog No. 17-987-113A
Explore available promotions
Description
Description
  • Urethane aprons designed for extended wear in food processing and life science industries.
  • Reliable protection against oil, grease, and chemicals.
  • Nylon reinforced to resist tearing, abrasion, and puncture.
  • Flexible lightweight construction.
  • Sealed grommets allow for size adjustment without tear-out.
  • Adjustable ties ensure optimal fit for wide range of wearers.
Specifications
Specifications
Bib Apron
Urethane
Reusable
Non-sterile
Sewn-edge
None
7 mil
48 x 35 in. (121.92 x 88.9 cm)
121.92 cm
88.9 cm
1 Ea.
Sewn
Medium
Yellow
Integral Ties
Nylon Reinforced Urethane, Sewn Edges
Yes
Tearing, Abrasion, Puncture
Food Service, Maintenance, Transferring Liquids
48 in.
35 in.
48/Cs.
Case