Ansul™ SPILL-X™ Agents Spill Treatment Kits
Mount easily on a wall yet remove quickly
Supplier:  Ansul™ 78774
 View more versions of this product
Catalog No. 17-987-144A
Explore available promotions
Includes: Spill response instructions, spill treatment guide, safety goggles, rubber gloves, disposal bags, scraper and pickup pans; pH paper included with all kits except for formaldehyde (17-987-144E).
Description
Description
Mount easily on a wall yet remove quickly
  • Six agents in shaker bottles plus additional spill response accessories.
  • Durable, bright green “suitcase” designed to be mounted easily on a wall and removed quickly
  • Multi-agent Kit contains two shaker bottles each of SPILL-X-A™, SPILL-X-C™ and SPILL-X-S™ agents
Order Info
Kits can be restocked.
Specifications
Specifications
6 agents in shaker bottles, guide, goggles, gloves, disposal bags, scraper, pH test papers, pick-up pans, green suitcase, Spill-X agents, 2 Shaker bottles/each Spill-X-A, Spill-X-C, Spill-X-S, instructions
Wall mount