Ansul™ SPILL-X™ Agents Spill Treatment Kits
Mount easily on a wall yet remove quickly
$1133.38 - $1463.93
Specifications
Mounting | Wall mount |
---|
Includes: Spill response instructions, spill treatment guide, safety goggles, rubber gloves, disposal bags, scraper and pickup pans; pH paper included with all kits except for formaldehyde (17-987-144E).
Catalog Number | Mfr. No. | Type | For Use With (Application) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | For Use With (Application) | Price | Quantity | ||||
17-987-144D
|
Ansul™
78778 |
Single Agent Spill Treatment Kit | Solvent |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
17-987-144A
|
Ansul™
78774 |
Each for $1,133.38
|
|
||||||
17-987-144C
|
Ansul™
78777 |
Single Agent Spill Treatment Kit | Caustic |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
17-987-144E
|
Ansul™
73834 |
Single Agent Spill Treatment Kit | Formaldehyde, solvent |
Each for $1,463.93
|
|
||||
Description
Mount easily on a wall yet remove quickly
Six agents in shaker bottles plus additional spill response accessories.
- Durable, bright green “suitcase” designed to be mounted easily on a wall and removed quickly
- Multi-agent Kit (78774) contains two shaker bottles each of SPILL-X-A™, SPILL-X-C™ and SPILL-X-S™ agents
Order Info
Kits can be restocked.
Specifications
Wall mount |