Ansul™ SPILL-X™ Agents Spill Treatment Kits
Mount easily on a wall yet remove quickly
Supplier:  Ansul™ 78777
 View more versions of this product
Catalog No. 17-987-144C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Includes: Spill response instructions, spill treatment guide, safety goggles, rubber gloves, disposal bags, scraper and pickup pans; pH paper included with all kits except for formaldehyde (17-987-144E).
Description
Description
Mount easily on a wall yet remove quickly
  • Six agents in shaker bottles plus additional spill response accessories.
  • Durable, bright green “suitcase” designed to be mounted easily on a wall and removed quickly
Order Info
Kits can be restocked.
Specifications
Specifications
Single Agent Spill Treatment Kit
Caustic
6 agents in shaker bottles, instructions, guide, goggles, gloves, disposal bags, scraper, pH test papers, pick-up pans, green suitcase