missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Anthranilamide, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 104900050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Anthranilamide | |
111.0°C to 114.0°C | |
198°C | |
99% min. (HPLC) | |
C7H8N2O | |
MFCD00007981 | |
14, 320 | |
Solubility in water: <5g/L water (20°C). Other solubilities: 140g/L ethanol (20°C), slightly soluble in ethylether, benzene | |
C1=CC=C(C(=C1)C(=O)N)N | |
136.15 | |
136.15 | |
Crystalline Powder or Flakes |
88-68-6 | |
300.0°C | |
Authentic | |
Glass bottle | |
2-(H2N)C6H4CONH2 | |
5 g | |
anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van | |
PXBFMLJZNCDSMP-UHFFFAOYSA-N | |
2-aminobenzamide | |
6942 | |
99+% |
Chemical Identifiers
88-68-6 | |
136.15 | |
PXBFMLJZNCDSMP-UHFFFAOYSA-N | |
6942 | |
C1=CC=C(C(=C1)C(=O)N)N |
C7H8N2O | |
MFCD00007981 | |
anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van | |
2-aminobenzamide |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 201-851-2
RUO – Research Use Only