missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Anthranilamide, 99+%, Thermo Scientific Chemicals
$56.17 - $71.45
Chemical Identifiers
CAS | 88-68-6 |
---|---|
Molecular Formula | C7H8N2O |
Molecular Weight (g/mol) | 136.15 |
MDL Number | MFCD00007981 |
InChI Key | PXBFMLJZNCDSMP-UHFFFAOYSA-N |
Synonym | anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van |
PubChem CID | 6942 |
IUPAC Name | 2-aminobenzamide |
SMILES | C1=CC=C(C(=C1)C(=O)N)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC104900050
|
Thermo Scientific Chemicals
104900050 |
5 g | Glass bottle |
Each for $56.17
|
|
||||
AC104901000
|
Thermo Scientific Chemicals
104901000 |
100 g | Plastic bottle |
Each for $71.45
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
88-68-6 | |
136.15 | |
PXBFMLJZNCDSMP-UHFFFAOYSA-N | |
6942 | |
C1=CC=C(C(=C1)C(=O)N)N |
C7H8N2O | |
MFCD00007981 | |
anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van | |
2-aminobenzamide |
Specifications
88-68-6 | |
300.0°C | |
Authentic | |
Glass bottle | |
2-(H2N)C6H4CONH2 | |
5 g | |
anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van | |
PXBFMLJZNCDSMP-UHFFFAOYSA-N | |
2-aminobenzamide | |
6942 | |
99+% | |
Anthranilamide |
111.0°C to 114.0°C | |
198°C | |
99% min. (HPLC) | |
C7H8N2O | |
MFCD00007981 | |
14, 320 | |
Solubility in water: <5g/L water (20°C). Other solubilities: 140g/L ethanol (20°C), slightly soluble in ethylether, benzene | |
C1=CC=C(C(=C1)C(=O)N)N | |
136.15 | |
136.15 | |
Crystalline Powder or Flakes |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 201-851-2
RUO – Research Use Only